Anti-FBXO28 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9088R
10666-866EA 796.88 CAD
10666-866
Anti-FBXO28 Rabbit Polyclonal Antibody
Antibodies
FBXO28 is a member of the F-box protein family. Such proteins are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: FBXO28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR