Anti-ANKS1B Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9097R
10667-004EA 796.88 CAD
10667-004
Anti-ANKS1B Rabbit Polyclonal Antibody
Antibodies
The β-Amyloid protein precursor (AbPP) is a widely expressed transmembrane protein that is processed into the b-Amyloid (Ab) peptide, which accumulates in insoluble plaques in the brain of Alzheimer’s disease patients and AbPP intracellular domain (AID). AID may function as a pro-apoptotic peptide, a regulator of calcium homeostasis and a molecule involved in transcriptional regulation. The AID associated protein 1 (AIDA-1) is highly expressed in the brain and is regulated by AbPP. It interacts with AbPP to play a role in brain development. AIDA-1 also interacts with coilin in Cajal bodies to regulate pre-mRNA splicing.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANKS1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR