Anti-ARSK Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9102R
10667-126EA 796.88 CAD
10667-126
Anti-ARSK Rabbit Polyclonal Antibody
Antibodies
Belongs to the sulfatase family. Sulfatases such as ARSK, hydrolyze sulfate esters from sulfated steroids,carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARSK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR