Anti-TAS1R3 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-9113R
10667-318EA 677.18 CAD
10667-318
Anti-TAS1R3 Rabbit Polyclonal Antibody
The TAS1R3 gene encodes the human homolog of mouse Sac, a major determinant of differences between sweet-sensitive and -insensitive mouse strains in their responsiveness to sucrose, saccharine, and other sweeteners (Max et al., 2001 [PubMed 11326277]).[supplied by OMIM, Jan 2010].

  • Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TAS1R3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR