Anti-FBXO38 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9122R
10667-504EA 796.88 CAD
10667-504
Anti-FBXO38 Rabbit Polyclonal Antibody
Antibodies
F box protein 38 probably recognizes and binds to some phosphorylated proteins and promotes their ubiquitination and degradation. It may coactivate KLF7, but does not seem to promote KLF7 ubiquitination. There are three named isoforms.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: FBXO38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR