Anti-ZNRF4 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9142R
10667-894EA 796.88 CAD
10667-894
Anti-ZNRF4 Rabbit Polyclonal Antibody
Antibodies
ZNRF4 contains 1 PA (protease associated) domain and 1 RING-type zinc finger.

Recommended Dilutions: Western Blot: 1:100-1000; Flow Cytometry: 1:20-100; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNRF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR