Anti-RNF87 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9166R
10668-312EA 796.88 CAD
10668-312
Anti-RNF87 Rabbit Polyclonal Antibody
Antibodies
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc binding domains, a RING, a B box type 1 and a B box type 2, and a coiled coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternative splicing of this gene generates three transcript variants, named alpha, beta and gamma.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF87
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR