Anti-Repetin Rabbit Polyclonal Antibody

BS-9181R
10668-616EA 435.03 CAD
10668-616
Anti-Repetin Rabbit Polyclonal Antibody
Repetin is a 784 amino acid secreted protein that belongs to the S100-fused protein family. Repetin is an extracellular epidermal matrix protein that is expressed in the epidermis and at high levels in eccrine sweat glands, the inner sheaths of hair roots and the filiform papilli of the tongue. A multifunctional epidermal matrix protein, repetin is involved in the cornified cell envelope formation. Containing two EF-hand domains, repetin’s recombinant N-terminal domain binds calcium reversibly. Repetin and trichohyalin may compensate for absent loricrin. The repetin gene contains three exons, two introns and spans at least 5.6 kb. The repetin gene is conserved in cow, mouse and rat, and maps to human chromosome 1q21.3.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Repetin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR