Anti-VGLL2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9183R
10672-126EA 796.88 CAD
10672-126
Anti-VGLL2 Rabbit Polyclonal Antibody
Antibodies
Vgl-2, also known as VITO-1, is a 317 amino acid protein that contains a domain through which it interacts with TEF-1, a protein that plays a role in controlling the expression of numerous genes. Specific to skeletal muscle, Vgl-2 is expressed highly in adult fast muscle and is expressed at lower levels in adult slow muscle and fetal skeletal muscle. During muscle differentiation, Vgl-2 mRNA levels increase and Vgl-2 translocates from the cytoplasm to the nucleus. Overexpression of Vgl-2 in MYOD-transfected 10T1/2 mouse embryonic fibroblasts increases expression of myosin heavy chain (MHC), which is a marker of terminal muscle differentiation. This evidence suggests that Vgl-2 is essential for muscle gene expression. There are two isoforms of Vgl-2 that are produced as a result of alternative splicing events.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: VGLL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR