Anti-IGSF6 Rabbit Polyclonal Antibody

BS-9198R
10668-924EA 407.07 CAD
10668-924
Anti-IGSF6 Rabbit Polyclonal Antibody
Ig (immunoglobulin) superfamily members exhibit functional characteristics including immune responses, growth factor signaling and cell adhesion. IGSF6 (immunoglobulin superfamily, member 6), also known as DORA, is a novel 241 amino acid single-pass type I membrane protein that contains one Ig-like C2-type (immunoglobulin-like) domain. Expressed in spleen, dendritic cells, peripheral blood lymphocytes and lymph node, IGSF6 is induced by TNF?and GM-CSF in dendritic cells and downregulated by ionomycin and PMA in monocytes. IGSF6 may function as a co-receptor in the antigen uptake complex or dendritic cell recirculation and is encoded by a gene located on human chromosome 16p12.2, a locus associated with inflammatory bowel disease.

Recommended Dilutions: Western Blot: 1:100-1000; Flow Cytometry: 1:20-100; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IGSF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR