Anti-SERPINB10 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9211R
10669-220EA 796.88 CAD
10669-220
Anti-SERPINB10 Rabbit Polyclonal Antibody
Antibodies
SERPINB10 is a serine proteinase inhibitor of the ovalbumin like B clade of serpins. It was first discovered in human bone marrow in a search for serpins similar to SerpinB6. SERPINB10 was identified in lymphocytes, and was elevated in chronic myeloid leukemia and chronic myelomonocytic leukemia cells. The monocytic cell lines THP-1 and AML-193 also showed elevated SERPINB10 expression.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SERPINB10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR