Anti-RNF180 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9233R
10669-628EA 796.88 CAD
10669-628
Anti-RNF180 Rabbit Polyclonal Antibody
Antibodies
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. RNF180 (ring finger protein 180), also known as Rines, is a 592 single-pass membrane protein that contains a single RING-type zinc finger. Expressed as three alternatively spliced isoforms, RNF180 is well conserved among vertebrates. RNF180 is expressed in brain, kidney, testis and uterus and localizes to the endoplasmic reticulum. RNF180 is an E3 ubiquitin ligase involved in the ubiquitin-proteasome pathway. RNF180 is encoded by a gene located on human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm, or of chromosome 5 altogether, is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF180
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR