Anti-RNF20 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9243R
10669-844EA 796.88 CAD
10669-844
Anti-RNF20 Rabbit Polyclonal Antibody
Antibodies
RNF20 is an E3 ubiquitin ligase protein that mediates monoubiquitination of histone H2B and the methylation of histone H3. It forms a ubiquitin ligase complex in cooperation with the E2 enzyme UBE2E1/UBCH6. It thereby plays a central role in histone code and gene regulation. It is required for transcriptional activation of Hox genes. It is recruited to the MDM2 promoter, probably by being recruited by p53/TP53, and thereby acts as a transcriptional coactivator.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR