Anti-MARCH10 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9245R
10672-322EA 796.88 CAD
10672-322
Anti-MARCH10 Rabbit Polyclonal Antibody
Antibodies
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. MARCH10 (membrane-associated ring finger (C3HC4) 10), also known as RNF190 (ring finger protein 190) or MARCH-X, is an 808 amino acid protein with one RING-CH-type zinc finger domain. MARCH10 may function as an E3 ubiquitin-protein ligase that accepts a ubiquitin residue from an E2 ubiquitin-conjugating enzyme and immediately transfers that residue to a protein that is targeted for degradation.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: MARCH10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR