Anti-RNF130 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9252R
10670-036EA 796.88 CAD
10670-036
Anti-RNF130 Rabbit Polyclonal Antibody
Antibodies
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. RNF130 (ring finger protein 130), also known as GP, G1RZFP (G1-related zinc finger protein) or GOLIATH, is a 419 amino acid single-pass type I membrane protein that shares similarity with a Drosophila zinc-finger protein found in mesoderm known as g1. RNF130 contains one PA (protease associated) domain and a single RING-type zinc finger. Implicated in the regulation of growth factor withdrawal-induced apoptosis of myeloid precursor cells, RNF130 is encoded by a gene located on human chromosome 5q35.3 and mouse chromosome 11 B1.3.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF130
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR