Anti-RNF133 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9253R
10670-060EA 796.86 CAD
10670-060
Anti-RNF133 Rabbit Polyclonal Antibody
Antibodies
RNF133 contains a RING finger domain, a motif known to be involved in protein-protein and protein-DNA interactions. This gene has no intron. RNF133 may have E3 ubiquitin-protein ligase activity.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF133
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR