Anti-RNF121 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9255R
10670-110EA 796.88 CAD
10670-110
Anti-RNF121 Rabbit Polyclonal Antibody
Antibodies
RNF121 protein contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported. Three of them are supported by at least two independent transcripts or ESTs, the full length natures of others are not clear.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF121
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR