Anti-RNF169 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9259R
10670-168EA 796.62 CAD
10670-168
Anti-RNF169 Rabbit Polyclonal Antibody
Antibodies
RNF169 contains 1 RING type zinc finger. The exact functions of RNF169 remain unknown.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF169
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR