Anti-BNC2 Rabbit Polyclonal Antibody

BS-9293R
10670-544EA 397.94 CAD
10670-544
Anti-BNC2 Rabbit Polyclonal Antibody
Probable transcription factor specific for skin keratinocytes. May play a role in the differentiation of spermatozoa and oocytes. There are two isoforms.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: BNC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR