Anti-ANKRD17 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9295R
10670-594EA 796.88 CAD
10670-594
Anti-ANKRD17 Rabbit Polyclonal Antibody
Antibodies
ANKRD17 is a 2,603 amino acid protein that contains 25 ankyrin repeats and one KH domain. ANKRD17 is expressed in bone marrow and is thought to be involved in liver development. ANKRD17 localizes to the cytoplasm and the nucleus. ANKRD17 exists as five alternatively spliced isoforms that are encoded by a gene which maps to human chromosome 4. Representing approximately 6% of the human genome, chromosome 4 contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease, is on chromosome 4. FGFR-3 is also encoded on chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer. Chromosome 4 is also tied to Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease. Chromosome 4 reportedly contains the largest gene deserts (regions of the genome with no protein encoding genes) and has one of the two lowest recombination frequencies of the human chromosomes.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANKRD17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR