Anti-BCL7B Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9298R
10486-580EA 796.88 CAD
10486-580
Anti-BCL7B Rabbit Polyclonal Antibody
Antibodies
BCL7B shows high homology to the BCL7A protein, which is known to be directly involved in a complex chromosomal translocation in Burkitt lymphoma cell lines. The specific function of BCL7B has not yet been determined. However, it may play a role in lung tumor development or progression. The BCL7B gene is located at a chromosomal region commonly deleted in the congenital disorder, Williams syndrome.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCL7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR