Anti-GPSM2 Rabbit Polyclonal Antibody

BS-9310R
10493-130EA 435.03 CAD
10493-130
Anti-GPSM2 Rabbit Polyclonal Antibody
Plays an important role in spindle pole orientation. Interacts and contributes to the functional activity of G(i) alpha proteins. Acts to stabilize the apical complex during neuroblast divisions.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPSM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR