Anti-NNF1R Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9319R
10495-118EA 796.88 CAD
10495-118
Anti-NNF1R Rabbit Polyclonal Antibody
Antibodies
NNF1R, also called PMF1, is part of the MIS12 complex, which may be fundamental for kinetochore formation and proper chromosome segregation during mitosis. It is required for chromosome congression and for correct operation of the spindle checkpoint. May act as a cotranscription partner of NFE2L2 involved in regulation of polyamine-induced transcription of SSAT.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NNF1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR