Anti-NYS48 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9320R
10486-692EA 796.88 CAD
10486-692
Anti-NYS48 Rabbit Polyclonal Antibody
Antibodies
NYS48 belongs to the HAUS8 family. It is required for stabilizing spindle integrity and chromosomal integrity.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NYS48
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR