Anti-MARCH6 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9340R
10486-862EA 796.62 CAD
10486-862
Anti-MARCH6 Rabbit Polyclonal Antibody
Antibodies
MARCH6 (Membrane associated RING finger protein 6) belongs to the MARCH family, which contains at least seven membrane associated RING-CH (MARCH)proteins. MARCH proteins are E3 ubiquitin ligases and are located to subcellular membranes.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: MARCH6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR