Anti-PSMA3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9357R
10487-088EA 796.88 CAD
10487-088
Anti-PSMA3 Rabbit Polyclonal Antibody
Antibodies
The proteasome is a multicatalytic proteinase complex with a highly ordered ring shaped 20S core structure. The core structure is composed of 4 rings of 28 non identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin dependent process in a non lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PSMA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR