Anti-PGBD1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9370R
10487-234EA 796.88 CAD
10487-234
Anti-PGBD1 Rabbit Polyclonal Antibody
Antibodies
PGDB1, also known as SCAND4 or HUCEP-4, is an 809 amino acid protein that contains one SCAN box domain and is a member of the PGBD family. Expressed specifically in brain tissue, PGBD1 may, via its SCAN box domain, be involved in transcriptional regulation events within the nucleus.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PGBD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR