Anti-CNOT6 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-9386R
10487-386EA 677.18 CAD
10487-386
Anti-CNOT6 Rabbit Polyclonal Antibody
CNOT6 is a widely expressed subunit of the CCR4-NOT transcription complex. The CCR4-NOT complex is an evolutionarily conserved, multi-component complex known to be involved in transcription as well as mRNA degradation. Various subunits (e.g. CNOT1, CNOT3) are involved in influencing nuclear hormone receptor activities. The CCR4-NOT complex is also involved in the regulation of Histone H3 lysine 4 methylation through a ubiquitin-dependent pathway that likely involves the proteasome. CNOT6 belongs to the CCR4/nocturin family and contains three LRR (leucine-rich) repeats. In the cytoplasm, CNOT6 acts as a poly(A) nuclease involved in mRNA decay mediated by the major-protein-coding determinant of instability (mCRD) of the Fos gene.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CNOT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR