Anti-LRSAM1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9387R
10489-446EA 796.88 CAD
10489-446
Anti-LRSAM1 Rabbit Polyclonal Antibody
Antibodies
LRSAM1 is an E3 ubiquitin-protein ligase that mediates monoubiquitination of TSG101 at multiple sites, leading to inactivation of the ability of TSG101 to sort endocytic (EGF receptors) and exocytic (HIV-1 viral proteins) cargos. It selectively regulates cell adhesion molecules and plays a role in receptor endocytosis and viral budding. LRSAM1 contains a RING-type zinc finger, 5 leucine-rich repeats and 1 SAM (sterile alpha motif) domain. The coiled coil domains interact with the SB domain of TSG101. The PTAP motifs mediate the binding to UEV domains. There are 3 isoforms produced by alternative splicing.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRSAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR