Anti-OGN Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9407R
10489-650EA 796.88 CAD
10489-650
Anti-OGN Rabbit Polyclonal Antibody
Antibodies
The small leucine-rich proteoglycan (SLRP) family of proteins contains various proteins such as Decorin, Biglycan, Fibromodulin, Keratocan, Lumican, Osteoadherin and Osteoglycin. These proteins all have similar functions as they all mediate extracellular matrix organization and act as binding partners of TGF beta. Osteoglycin, which also may be designated osteoinductive factor (OIF), is a secreted protein detected in bone tissues. Osteoglycin induces the formation of bone in conjunction with either TGF-beta-1 or TGF-beta-2. The precursor form of the OGN gene product, designated Mimecan, is subject to in situ proteolytic cleavage to yield the mature Osteoglycin.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: OGN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR