Anti-TSPAN2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9411R
10495-320EA 796.88 CAD
10495-320
Anti-TSPAN2 Rabbit Polyclonal Antibody
Antibodies
TSPAN2 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. TSPAN2 may play a role in signalling in oligodendrocytes in the early stages of their terminal differentiation into myelin-forming glia and may also function in stabilizing the mature sheath.

Recommended Dilutions: Western Blot: 1:100-1000; Flow Cytometry: 1:20-100; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSPAN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR