Anti-TSPAN3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9412R
10487-722EA 796.62 CAD
10487-722
Anti-TSPAN3 Rabbit Polyclonal Antibody
Antibodies
TSPAN3 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene. Two alternative transcripts encoding different isoforms have been described.

Recommended Dilutions: Western Blot: 1:100-1000; Flow Cytometry: 1:20-100; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSPAN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR