Anti-TSPAN6 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9416R
10487-802EA 796.88 CAD
10487-802
Anti-TSPAN6 Rabbit Polyclonal Antibody
Antibodies
Tetraspanins are a group of hydrophobic membrane proteins that interact with a wide variety of proteins including intracellular signaling molecules, integrins and membrane receptors. TSPAN7 (tetraspanin 7), also known as MXS1 (membrane component chromosome X surface marker 1) or TM4SF2 (transmembrane 4 superfamily member 2), is a 249 amino acid multi-pass membrane protein belonging to the tetraspanin (TM4SF) family of transmembrane proteins. TSPAN7 is believed to play a role in cell motility and cell proliferation. The gene that encodes TSPAN7 maps to human chromosome X and defects in this gene are a cause of mental retardation X-linked type 58 (MRX58), which is characterized by dramatically below average general intellectual functioning.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSPAN6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR