Anti-TSPAN16 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9423R
10487-864EA 796.88 CAD
10487-864
Anti-TSPAN16 Rabbit Polyclonal Antibody
Antibodies
TM4-B, also known as TSPAN16 (tetraspanin-16) or TM4SF16 (transmembrane 4 superfamily member 16), is a 245 amino acid multi-pass membrane protein that belongs to the tetraspanin (TM4SF) family. Members of the TM4SF family mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. TM4-B is broadly expressed in most human tissues and cell lines including neural and bone marrow derived tissues. The gene that encodes TM4-B contains nearly 31,000 bases and maps to human chromosome 19p13.2. Chromosome 19 consists of over 63 million bases, houses approximately 1,400 genes and is recognized for having the greatest gene density of the human chromosomes. It is the genetic home for a number of immunoglobulin (Ig) superfamily members, including the killer cell and leukocyte Ig-like receptors, a number of ICAMs, the CEACAM and PSG families and Fc receptors (FcRs).

Recommended Dilutions: Western Blot: 1:100-1000; Flow Cytometry: 1:20-100; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSPAN16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR