Anti-TSPAN15 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9425R
10489-790EA 796.89 CAD
10489-790
Anti-TSPAN15 Rabbit Polyclonal Antibody
Antibodies
NET-7, also known as TSPAN15 (tetraspanin 15) or TM4SF15 (transmembrane 4 superfamily member 15), is a 294 amino acid multi-pass membrane protein that belongs to the transmembrane 4 superfamily, also known as the tetraspanin family. Members of the tetraspanin family are cell-surface proteins that are characterized by the presence of four hydrophobic domains and mediate signal transduction events that play a role in the regulation of cell development, activation, growth, motility, differentiation, and cancer. Considered molecular facilitators, tetraspanin proteins may regulate vesicle fusion and fission.

Recommended Dilutions: Western Blot: 1:100-1000; Flow Cytometry: 1:20-100; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSPAN15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR