Anti-PYGB Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9453R
10488-128EA 796.88 CAD
10488-128
Anti-PYGB Rabbit Polyclonal Antibody
Antibodies
Glycolysis is an evolutionarily conserved series of ten chemical reactions that utilizes eleven enzymes to concomitantly generate pyruvate and ATP from glucose. fructose kinase-2/fructose 2,6-bisphosphatase (PFK-2) stimulates the synthesis and degradation of fructose 2,6-bisphosphate. Glycogen phosphorylase (also known as GP) is an allosteric enzyme important in carbohydrate metabolism. Its activity is regulated through either noncovalent binding of metabolites or by covalent modification. Glycogen phosphorylase catalyzes the phosphorylation of glycogen to Glc-1-P. There are three genes which encode the brain, liver and muscle forms of glycogen phosphorylase, PYGB, PYGL and PYGM. Because of its fundamental role in the metabolism of glycogen, glycogen phosphorylase has been a target for the design of inhibitory compounds, which could be valuable in the therapeutic treatment of type 2 diabetes mellitus.

Recommended Dilutions: IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PYGB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR