Anti-PROCR Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9506R
10488-994EA 796.86 CAD
10488-994
Anti-PROCR Rabbit Polyclonal Antibody
Antibodies
EPCR is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. It is an N glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregnancy. It is expressed strongly in the endothelial cells of arteries and veins in heart and lung. It is overexpressed in several human non-Pgp multidrug resistant (MDR) tumor cell lines. Whether and how anti-coagulant and anti-inflammatory functions of EPCR facilitate oncogenesis and/or drug resistance is unclear.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: PROCR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR