Anti-IL15 Rabbit Polyclonal Antibody

BS-9565R
10491-586EA 397.94 CAD
10491-586
Anti-IL15 Rabbit Polyclonal Antibody
Neurotransmission depends on the regulated exocytotic release of chemical transmitter molecules. This requires the packaging of these substances into the specialized secretory vesicles of neurons and neuroendocrine cells, a process mediated by specific vesicular transporters. The family of genes encoding the vesicular transporters of monoamines (VMAT 1 and VMAT 2) and acetylcholine (VACht) have been cloned and functionally characterized. The sequence of these integral membrane proteins predicts twelve transmembrane domains and weak homology to a class of bacterial antibiotic resistance proteins. The vesicular transport of neurotransmitter molecules has been shown to be an active ATP- and proton dependent transport mechanism.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR