Anti-TSPEAR Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9577R
10491-766EA 796.88 CAD
10491-766
Anti-TSPEAR Rabbit Polyclonal Antibody
Antibodies
TSPEAR, also known as C21orf9, is a 669 amino acid secreted protein. Expressed as two isoforms produced by alternative splicing, TSPEAR contains one Thrombospondin N-terminal domain and seven EAR (epilepsy-associated repeat) domains. EAR domains are found in several proteins, including TSPEAR, encoded by genes that map within chromosome regions associated with seizure disorders. It is thought that the EAR domain plays a role in the pathogenesis of epilepsy by either binding to an unknown epileptic ligand or interfering with axon synaptogenesis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSPEAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR