Anti-DRIP1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9618R
10493-458EA 796.88 CAD
10493-458
Anti-DRIP1 Rabbit Polyclonal Antibody
Antibodies
1.C14ORF28, GNB2L1, MLLT3, DRD2 and DARPP-32 are important in the pathogenesis of schizophrenia and bipolar disorder; 2. these two disorders share common disease-related mechanisms linked to dopamine signalling; 3. the expression of these genes is closely correlated; and 4. DRD2 provides the initial trigger in the pathogenesis of these disorders.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: DRIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR