Anti-RBFA Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9653R
10494-012EA 796.88 CAD
10494-012
Anti-RBFA Rabbit Polyclonal Antibody
Antibodies
C18orf22, also known as HsT169, is a 343 amino acid protein that localizes to the mitochondrion and is expressed as two alternatively spliced isoforms that are encoded by a gene which maps to human chromosome 18. Chromosome 18 houses over 300 protein-coding genes and contains nearly 76 million nucleotide bases. There are a variety of diseases associated with defects in chromosome 18-localized genes, some of which include Trisomy 18 (also known as Edwards syndrome), Niemann-Pick disease, hereditary hemorrhagic telangiectasia, erythropoietic protoporphyria and follicular lymphomas.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: RBFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR