Anti-FAM29A Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9723R
10496-158EA 796.88 CAD
10496-158
Anti-FAM29A Rabbit Polyclonal Antibody
Antibodies
The human augmin complex (HAUS) is an evolutionarily conserved 8-subunit protein complex that was initially discovered in Drosophila. The HAUS complex is essential for microtubule generation, centrosome integrity, mitotic spindle assembly and completion of cytokinesis. HAUS6 (HAUS augmin-like complex, subunit 6), also known as FAM29A or Dgt6, is a 955 amino acid component of the augmin complex. Required for mitotic progression, HAUS6 localizes to cytoplasm, cytoskeleton, mitotic spindle microtubules and interphase centrosomes, and undergoes post-translational phosphorylation following mitosis on multiple serine and threonine residues. HAUS6 exists as two alternatively spliced isoforms that are encoded by a gene located on human chromosome 9.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM29A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR