Anti-RMDN1 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9730R
10496-232EA 796.88 CAD
10496-232
Anti-RMDN1 Rabbit Polyclonal Antibody
Antibodies
FAM82B belongs to the FAM82/RMD family and contains two TPR repeats. The specific function of FAM82B is not yet known.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: RMDN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR