Anti-EML3 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-9731R
10496-256EA 796.88 CAD
10496-256
Anti-EML3 Rabbit Polyclonal Antibody
Antibodies
At the onset of mitosis, assembly of the mitotic spindle requires a global change in the activity of microtubule-binding proteins. EML3 (Echinoderm microtubule-associated protein-like 3) is a 896 amino acid protein that likely modifies microtubule dynamics by making them longer. Through colocalization with spindle microtubules during mitosis, EML3 plays a role in correct metaphase chromosome alignment. EML3 contains a nuclear localization signal and a microtubule-binding domain. The gene encoding EML3 maps to human chromosome 11, which houses over 1,400 genes and comprises nearly 4% of the human genome. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are associated with defects in genes that maps to chromosome 11.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: EML3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR