H-D-Arg(Me)-Oh
Supplier: Bachem
4082009.0025 4082009.0050 4082009.0100
F-4275.0025BAEA 215.45 CAD
F-4275.0025BA F-4275.0050BA F-4275.0100BA
H-D-Arg(Me)-Oh
Negative control for L-NMMA and ADMA.CAS Number (net): 137694-74-7.
Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.