Human Recombinant FGF-9 (from E. coli)
Supplier: PeproTech
100-23-500UG 100-23-20UG 100-23-100UG 100-23-1MG 100-23-5UG 100-23-250UG
10772-088EA 4498.02 CAD
10772-088 10772-082 10772-084 10772-090 10772-080 10772-086
Human Recombinant FGF-9 (from E. coli)
Proteins and Peptides
FGF-9 is a heparin-binding growth factor that belongs to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-9 targets glial cells, astrocytes cells and other cells that express the FGFR 1c, 2c, 3b, 3c, and 4. Recombinant Human FGF-9 is a 23.2 kDa protein consisting of 206 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.