Human Recombinant FGF-5 (from E. coli)
Supplier: PeproTech
100-34-10UG 100-34-1MG 100-34-50UG 100-34-500UG 100-34-100UG 100-34-250UG
10772-190EA 150.18 CAD
10772-190 10772-200 10772-192 10772-198 10772-194 10772-196
Human Recombinant FGF-5 (from E. coli)
Proteins and Peptides
FGF-5 is a secreted, heparin-binding growth factor that belongs to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-5 binds to FGFR 1c and 2c, and plays a regulatory role in the hair growth cycle. Recombinant Human FGF-5 is a 27.6 kDa protein consisting of 252 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.