Human GDF15 (from Cells)
Supplier: PeproTech
120-28-20UG 120-28-5UG 120-28-500UG 120-28-100UG 120-28-1MG
10772-650EA 366.09 CAD
10772-650 10772-648 10772-654 10772-652 10772-656
Human GDF15 (from Cells)
Proteins and Peptides
GDF-15 belongs to the TGF-β cytokine family, whose members play an important role during prenatal development and postnatal growth, and the remodeling and maintenance of a variety of tissues and organs. GDF-15 is expressed predominantly in the placenta and, to a much lesser extent, in various other tissues. The presence of GDF-15 in amniotic fluid and its elevated levels in the sera of pregnant women suggest GDF-15's involvement in gestation and embryonic development. GDF-15 generally exerts tumor suppressive activities and is one of the predominant factors produced and secreted in response to activation of the p53 pathway. Interestingly, the serum level of GDF-15 is positively correlated with neoplastic progression of several tumor types, including certain colorectal, pancreatic, and prostate cancers. Human GDF-15/MIC-1 is a disulfide linked homodimeric protein consisting of two 112 amino acid polypeptide chains. The calculated molecular weight of Human GDF-15/MIC-1 is 24.6 kDa.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.