Human Recombinant IL-1alpha (from E. coli)
Supplier: PeproTech
200-01A-50UG 200-01A-2UG 200-01A-100UG 200-01A-10UG 200-01A-250UG
10772-948EA 934.29 CAD
10772-948 10772-944 10772-950 10772-946 10772-952
Human Recombinant IL-1alpha (from E. coli)
Proteins and Peptides
IL-1α is a non-secreted, proinflammatory cytokine produced in a variety of cells, including monocytes, tissue macrophages, keratinocytes, and other epithelial cells. Both IL-1α and IL-1β bind to the same receptor and have similar, if not identical, biological properties. These cytokines have a broad range of activities including the stimulation of thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, mitogenic FGF-like activity and the release of prostaglandin and collagenase from synovial cells. However, whereas IL-1β is a secreted cytokine, IL-1α is predominantly a cell-associated cytokine. Recombinant Human IL-1α is an 18.0 kDa protein containing 159 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.