Human Recombinant IL-12P40 (from CHO cells)
Supplier: PeproTech
20012P40500UG 20012P40250UG 200-12P40-2UG 200-12P40-10UG 20012P40100UG 200-12P40-1MG
10773-032EA 8783.88 CAD
10773-032 10773-030 10773-024 10773-026 10773-028 10773-034
Human Recombinant IL-12P40 (from CHO cells)
Proteins and Peptides
IL-12 is a disulfide-linked heterodimeric protein (p70), composed of two subunits, p35 and p40, which are encoded by two different genes. Accumulating data indicates that p40 secretion precedes that of IL-12 expression. In addition, to its ability to covalently bind to p35 to form IL-12, p40 can bind to p19 to form IL-23. Recombinant Human IL-12 p40 is a 40.0 kDa protein containing 306 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.